Learn More
Description
Includes
Mouse MIP-1β; Standard; Mouse MIP-1β; PE Detection Reagent; Mouse MIP-1β; Capture Bead C9
The BD CBA Mouse MIP-1β Flex Set is a bead-based immunoassay capable of measuring mouse Macrophage Inflammatory Protein-1β (MIP-1β), also known as CCL4, in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse MIP-1β Flex Set.
- Reactivity: Mouse
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | MIP-1beta |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Mouse MIP-1beta, standard, Mouse MIP-1beta, PE detection reagent, Mouse MIP-1beta, capture bead C9 |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse MIP-1beta flex set (bead C9) |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.