Learn More
Description
Includes
Mouse RANTES; Standard; Mouse RANTES; PE Detection Reagent; Mouse RANTES; Capture Bead D8
Mouse reactivity is determined by testing samples with the BD CBA Mouse RANTES Flex Set.
Specifications
Specifications
| Antigen | RANTES |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Mouse RANTES, standard, Mouse RANTES, PE detection reagent, Mouse RANTES, capture bead D8 |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse RANTES flex set (bead D8) |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.