Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse Th1/Th2/Th17 Cytokine standards

Catalog No. bdb561665 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561665 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561665 Supplier BD Supplier No. 561665
Only null left
Add to Cart
Add to Cart

Designed for use as standard controls with the BD Cytometric Bead Array (CBA) Mouse Th1/Th2/TH17 Cytokine Kit

  • Contains a mixture of individually lyophilized recombinant proteins made up of recombinant Mouse Interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF)
  • Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Mouse
Includes Mouse interleukin (IL)-2, IL-4, IL-6, IL-10, IL-17A, interferon (IFN) and tumor necrosis factor (TNF)
Quantity 1 Vial
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry, Immunoassay
Product Type Mouse Th1/Th2/Th17 Cytokine standard

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.