Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Mouse TNF; Flex Set, Bead C8

Catalog No. BDB558299 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558299 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558299 Supplier BD Supplier No. 558299
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Mouse TNF; Standard; Mouse TNF; PE Detection Reagent; Mouse TNF; Capture Bead C8

For detection of mouse tumor necrosis factor in serum and cell culture supernatant samples

The BD CBA Mouse TNF Flex Set is a bead-based immunoassay capable of measuring mouse tumor necrosis factor (TNF) in serum and cell culture supernatant samples. Mouse and rat reactivity was determined by testing samples with the BD CBA Mouse TNF Flex Set. For quantitation of rat TNF, we recommend the BD™ CBA Rat TNF Flex Set (Cat. No. 558309).

  • Reactivity: Mouse and Rat
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Applications:

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Conjugate PE
Cross Reactivity Mouse
Includes Mouse TNF, standard, Mouse TNF, PE detection reagent, Mouse TNF, capture bead C8
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
Form Liquid
For Use With (Application) Flow Cytometry
Product Type Mouse TNF flex set (bead C8)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.