Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Phospho BLNK (Y84); Flex Set, Bead C9

Catalog No. BDB560063 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560063 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560063 Supplier BD Supplier No. 560063

Includes

Phospho BLNK (Y84); Standard; Phospho BLNK (Y84); PE Detection Reagent; Phospho BLNK (Y84); Capture Bead C9

Capable of measuring human B cell linker protein (BLNK) that has been tyrosine-phosphorylated at Tyr-84 in denatured cell lysate samples

  • Human reactivity was determined by testing cell lysates with the BD CBA Phospho BLNK (Y84) Flex Set

TRUSTED_SUSTAINABILITY

Specifications

Antigen BLNK (Y84)
Cross Reactivity Human
Includes Phospho BLNK (Y84), standard, Phospho BLNK (Y84), PE detection reagent, Phospho BLNK (Y84), capture bead C9
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Phospho BLNK (Y84) flex set (bead C9)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.