Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Phospho BTK(Y551); Flex Set, Bead D5

Catalog No. BDB560004 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560004 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560004 Supplier BD Supplier No. 560004

Includes

Phospho BTK(Y551), standard (one vial), Phospho BTK(Y551) PE Detection Reagent (one vial), Phospho BTK(Y551) Capture Bead D5 (one vial)

Bead-based immunoassay for measuring human Bruton's tyrosine kinase (Btk) which has been tyrosine-phosphorylated at Tyr-551 in denatured cell lysate samples

  • Human reactivity was determined by testing cell lysates with Flex Set
  • Theoretical limit of detection is 1.88 units/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen BTK(Y551)
Cross Reactivity Human
Includes Phospho BTK(Y551), standard (one vial), Phospho BTK(Y551) PE detection reagent (one vial), Phospho BTK(Y551) capture bead D5 (one vial)
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Phospho BTK (Y551) flex set (bead D5)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.