Learn More
BD Cytometric Bead Array (CBA) Phospho c-Jun (S63); Flex Set, Bead D8
Shop All BD Biosciences ProductsDescription
Includes
Phospho c-Jun (S63); Standard; Phospho c-Jun (S63); PE Detection Reagent; Phospho c-Jun (S63); Capture Bead D8
- Capable of measuring human c-Jun, a component of the activator protein-1 transcription factor, that has been serine-phosphorylated at Ser-63 in denatured cell lysate samples
Specifications
Specifications
| Antigen | c-Jun (S63) |
| Cross Reactivity | Human |
| Includes | Phospho c-Jun (S63), standard, Phospho c-Jun (S63), PE detection reagent, Phospho c-Jun (S63), capture bead D8 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Phospho c-Jun (S63) flex set ( bead D8) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.