Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Phospho eNOS; Flex Set, Bead C7

Catalog No. BDB560021 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560021 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560021 Supplier BD Supplier No. 560021

Includes

Phospho eNOS; Standard; Phospho eNOS; PE Detection Reagent; Phospho eNOS; Capture Bead C7

Bead-based immunoassay capable of measuring human endothelial nitric oxide synthase (eNOS), a cell-type specific enzyme that catalyzes the synthesis of nitric oxide, which has been serine-phosphorylated at Ser-1177 in denatured cell lysate samples

Human reactivity was determined by testing cell lysates with the BD CBA Phospho eNOS (S1177) Flex Set.

TRUSTED_SUSTAINABILITY

Specifications

Antigen eNOS
Cross Reactivity Human
Includes Capture bead, detection reagent and standard
Quantity 100 Tests
For Use With (Application) Flow Cytometry
Product Type Phospho eNOS (S1177) flex set (bead C7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.