Learn More
BD Cytometric Bead Array (CBA) Phospho ITK(Y511); Flex Set, Bead C6
Shop All BD Biosciences ProductsDescription
Includes
Phospho ITK(Y511), Standard (one vial), Phospho ITK(Y511) PE Detection Reagent (one vial), Phospho ITK(Y511) Capture Bead C6 (one vial)
- Human/mouse reactivity was determined by testing cell lysates Flex Set
- Theoretical limit of detection is 0.47units/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow cytometry
Specifications
Specifications
| Antigen | ITK(Y511) |
| Cross Reactivity | Human, Mouse |
| Includes | Phospho ITK(Y511), standard (one vial), Phospho ITK(Y511) PE detection reagent (one vial), Phospho ITK(Y511) capture bead C6 (one vial) |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Phospho ITK (Y511) flex set (bead C6) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.