Learn More
Description
Includes
Phospho JNK1/2; Standard; Phospho JNK1/2; PE Detection Reagent; Phospho JNK1/2; Capture Bead B5
- Capable of measuring human, mouse, or rat c-Jun N-terminal kinase1/2 (JNK1/2), that has been threonine-phosphorylated at Thr-183 and tyrosine-phosphorylated at Tyr-185 in denatured cell lysate samples
- Human and mouse reactivity was determined by testing cell lysates with the BD CBA Phospho JNK1/2 (T183/Y185) Flex Set
- Provides sufficient reagents for 100 tests
Order Info
Must be used in conjunction with a BD CBA Cell Signaling Master Buffer Kit (Cat. No. BDB560005, 100 tests, or BDB560006, 500 tests) and a flow cytometer.
Specifications
Specifications
| Antigen | JNK1/2 |
| Conjugate | PE |
| Cross Reactivity | Human, Mouse, Rat |
| Includes | Phospho JNK1/2, standard, Phospho JNK1/2, PE detection reagent, Phospho JNK1/2, capture bead B5 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Phospho JNK1/2 (T183/Y185) flex set (bead B5) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.