Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Phospho P38 Flex Set, Bead B6

Catalog No. BDB560010 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560010 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560010 Supplier BD Supplier No. 560010
Add to Cart
Add to Cart

Includes

Phospho P38; Standard; Phospho P38; PE Detection Reagent; Phospho P38; Capture Bead B6

A bead-based immunoassay

  • Capable of measuring human, mouse, or rat p38, a member of the mitogen-activated protein kinase (MAPK) family of kinases, that has been threonine-phosphorylated at Thr-180 and tyrosine-phosphorylated at Tyr-182 in denatured cell lysate samples
  • Human and mouse reactivity was determined by testing cell lysates with the BD CBA Phospho p38 (T180/Y182) Flex Set
  • Provides sufficient reagents for 100 tests

Order Info

Must be used in conjunction with a BD CBA Cell Signaling Master Buffer Kit (Cat. No. BDB560005, 100 tests, or BDB560006, 500 tests) and a flow cytometer.

TRUSTED_SUSTAINABILITY

Specifications

Antigen P38
Cross Reactivity Human, Mouse, Rat
Includes Phospho P38, standard, Phospho P38, PE detection reagent, Phospho P38, capture bead B6
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Phospho p38 (T180/Y182) flex set (bead B6)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.