Learn More
BD Cytometric Bead Array (CBA) Phospho SLP-76 (Y145); Flex Set, Bead D6
Shop All BD Biosciences ProductsDescription
Includes
Phospho SLP-76 (Y145); Standard; Phospho SLP-76 (Y145); PE Detection Reagent; Phospho SLP-76 (Y145); Capture Bead D6
- Capable of measuring human SH2 domain-containing Leukocyte Protein (SLP-76) that has been tyrosine-phosphorylated at Tyr-145 in denatured cell lysate samples
Specifications
Specifications
| Antigen | SLP-76 (Y145) |
| Cross Reactivity | Human |
| Includes | 1 Vial capture bead, 1 vial detection reagent and 2 vials standard |
| Quantity | 100 Tests |
| Sample Type | Denatured Cell Lysate |
| Content And Storage | 4°C |
| For Use With (Application) | Flow cytometry and measuring Human SH2 domain-containing Leukocyte Protein (SLP-76) that has been Tyrosine-Phosphorylated at Tyr-145 |
| Product Type | Phospho SLP-76 (Y145) flex set (bead D6) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.