Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Phospho STAT1 (Y701); Flex Set, Bead C5

Catalog No. BDB560015 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560015 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560015 Supplier BD Supplier No. 560015

Includes

Phospho STAT1 (Y701), Standard (one vial), Phospho STAT1 (Y701) PE Detection Reagent (one vial), Phospho STAT1 (Y701) Capture Bead C5 (one vial)

Bead-based immunoassay for measuring human or mouse signal transducer and activator of transcription 1 (Stat1) that has been tyrosine-phosphorylated at Tyr-701 in denatured cell lysate samples

  • Human and mouse reactivity was determined by testing cell lysates with Flex Set
  • Theoretical limit of detection is 1.36units/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen STAT1 (Y701)
Cross Reactivity Human
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard
Quantity 100 Tests
Sample Type Denatured Cell Lysate
Content And Storage 4°C
For Use With (Application) Flow cytometry and for measuring Human or Mouse signal transducer and activator of transcription 1 (Stat1) that has been Tyrosine-Phosphorylated at Tyr-701
Product Type Phospho Stat1 (Y701) flex set (bead C5)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.