Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Rat IFN-γ; Flex Set, Bead A6

Catalog No. BDB558305 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558305 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558305 Supplier BD Supplier No. 558305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Rat IFN-γ; Standard; Rat IFN-γ; PE Detection Reagent; Rat IFN-γ; Capture Bead A6

For detection of rat interferon in serum and cell culture supernatant samples

The BD CBA Rat IFN-γ Flex Set is a bead-based immunoassay capable of measuring rat interferon-γ (IFN-γ) in serum and cell culture supernatant samples. Rat reactivity was determined by testing samples with the BD CBA Rat IFN-γ Flex Set.

  • Reactivity: Rat
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-gamma
Cross Reactivity Rat
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow cytometry and for detection of Rat interferon
Product Type Rat IFN-gamma Flex set (bead A6)

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.