Learn More
Description
Includes
RAT IL-1α; Standard; RAT IL-1α; PE Detection Reagent; RAT IL-1α; Capture Bead A4
The BD CBA RAT IL-1α Flex Set is a bead-based immunoassay capable of measuring rat interleukin-1α (IL-1α) in serum and cell culture supernatant samples.
- Reactivity: QC Tested: Rat
- Bead Position: A4
- Limit of detection: 4.0pg/mL
With BD CBA Mouse/Rat Soluble Protein Master Buffer Kit (Cat. No. 558266, 100 tests, or 558267, 500 tests), a flow cytometer, and the FCAP Array™ software
Specifications
Specifications
| Antigen | IL-1alpha |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent and 2 vials standard |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Rat IL-1alpha flex set (bead A4) |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.