Learn More
BD Cytometric Bead Array (CBA) Rat IL-2 Enhanced Sensitivity Flex Set (Bead E5)
Shop All BD Biosciences ProductsDescription
Includes
- Rat IL-2 Standard
- Rat IL-2 PE Protection Reagent
- Rat IL-2 Capture Bead E5
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
- Protein concentration of 10,000 pg/ml (when reconstituted in 0.4 ml Assay Diluent)
Flow Cytometry
Specifications
Specifications
| Antigen | IL-2 |
| Conjugate | PE |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent and 2 vials standard |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Measures Rat interleukin-2 (IL-2) |
| Product Type | Rat IL-2 flex set (bead E5) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.