Learn More
Description
Includes
Rat TNF; Standard; Rat TNF; PE Detection Reagent; Rat TNF; Capture Bead C8
The BD CBA Rat TNF Flex Set is a bead-based immunoassay capable of measuring rat tumor necrosis factor (TNF) in serum and cell culture supernatant samples. Rat and mouse reactivity was determined by testing samples with the BD CBA Rat TNF Flex Set.
- Reactivity: Rat
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 10000pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
Specifications
| Antigen | TNF |
| Conjugate | PE |
| Cross Reactivity | Rat |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Sample |
| Content And Storage | 4°C |
| Form | Solid |
| For Use With (Application) | Flow cytometry, for detecting Rat tumor necrosis factor |
| Product Type | Rat TNF flex set (bead C8) |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.