Learn More
Description
Includes
- Total p38α Capture Bead B6
- Total p38-α PE Detection Reagent
- Total p38-α Standard
Contains one vial of each component and all components of have been formulated to a 50x concentration to ensure product performance when multiplexed. Lyophilized and the standard sphere should be transferred to 1.5mL microfuge tube for
reconstitution.
- Human reactivity was determined by testing cell lysates with this Flex set
- Must be used in conjunction with CBA Cell Signaling Master Buffer Kit and flow cytometer
Specifications
Specifications
| Antigen | p38-alpha |
| Cross Reactivity | Human |
| Includes | 1 Vial capture bead, 1 vial detection reagent and 2 vials standard |
| Quantity | 100 Tests |
| Sample Type | Cell Lysate |
| Content And Storage | 4°C |
| For Use With (Application) | Flow Cytometry |
| Product Type | Total p38-a flex set (bead B6) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.