Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Total ZAP70; Flex Set, Bead B8

Catalog No. BDB560019 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560019 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560019 Supplier BD Supplier No. 560019

Includes

Total ZAP70; Standard; Total ZAP70; PE Detection Reagent; Total ZAP70; Capture Bead B8

Bead-based immunoassay capable of measuring human, mouse or rat ZAP70

  • Human reactivity was determined by testing cell lysates with the BD CBA Total ZAP70 Flex Set
  • Reactivity with mouse and rat samples was determined by western blot for each of the antibodies used in the BD CBA Total ZAP70 Flex Set

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70
Cross Reactivity Human, Mouse, Rat
Includes 1 Vial capture bead, 1 vial detection reagent, 2 vials standard
Quantity 100 Tests
Sample Type Cell Lysate
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Total ZAP-70 flex set (bead B8)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.