Learn More
Description
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
- Concentration of 2500pg/mL when reconstituted in 4.0 mL Assay Diluent
- Recombinant
- Regulatory status: RUO
Flow cytometry
Order Info
One vial.
Specifications
Specifications
| Antigen | IL-12B, IL-12A |
| Cross Reactivity | Human |
| Quantity | 1 Vial |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human IL-12 p70 flex set standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.