Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array Flex Set Standard

Catalog No. BDB558456 Shop All BD Biosciences Products
Change view
Click to view available options
Description:
Human IFN-gamma Flex Set Standard
Human IL-10 Flex Set Standard
Human IL-1beta Flex Set Standard
Human IL-2 Flex Set Standard
Human IL-4 Flex Set Standard
Human IL-6 Flex Set Standard
Human IL-8 Flex Set Standard
Human TNF Flex Set Standard
Mouse IL-10 Flex Set Standard
Mouse IL-6 Flex Set Standard
Mouse TNF Flex Set Standard
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
BDB558456 Human IFN-gamma Flex Set Standard
BDB558457 Human IL-1beta Flex Set Standard
BDB558464 Human IL-6 Flex Set Standard
BDB558462 Human IL-4 Flex Set Standard
BDB558461 Human IL-2 Flex Set Standard
BDB558458 Human IL-10 Flex Set Standard
BDB558472 Human TNF Flex Set Standard
BDB558465 Human IL-8 Flex Set Standard
BDB558474 Mouse IL-10 Flex Set Standard
BDB558479 Mouse IL-6 Flex Set Standard
BDB558480 Mouse TNF Flex Set Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. BDB558456 Supplier BD Supplier No. 558456
Add to Cart
Add to Cart

Used as standard control

This product contains lyophilized recombinant human Interferon-λ (IFN-λ)

  • Storage Buffer: Lyophilized in aqueous buffered solution containing BSA and ProClin™ 150

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cytometric Bead Array (CBA) Standard
Description Human IFN-gamma Flex Set Standard
Content And Storage 4°C
Format Lyophilized
Includes Lyophilized recombinant Human Interferon-l (IFN-l)
Quantity 0.2 mL
Regulatory Status RUO
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.