Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array Mouse IFN-γ Flex Set Standard

Catalog No. BDB558473 Shop All BD Biosciences Products
Change view
Click to view available options
Kit Size:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Kit Size
BDB558473 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558473 Supplier BD Supplier No. 558473

missing translation for 'validMainframeMsg'

Useful as a standard control with CBA Mouse IFN-γ Flex Set

  • Contains lyophilized recombinant mouse interferon-γ (IFN-γ)
  • When reconstituted in 0.4mL assay diluent, has a protein concentration of 25,000pg/mL

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffered Solution containing BSA and ProClin™ 150
Includes Technical data sheet
Kit Size 100 Tests
Product Type Cytometric Ab Control Standards
For Use With (Application) Flow cytometry
Regulatory Status RUO
Protein Concentration 50000pg/mL
Storage Requirements 4°C
Volume (Metric) 0.2mL
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.