Learn More
BD Cytometric Bead Array Mouse IL-12p70 Enhanced Sensitivity Flex Set
Shop All BD Biosciences ProductsDescription
Contains one vial each of Enhanced Sensitivity Capture Bead and Detection Reagent (Part A) and two vials of Standard. The Enhanced Sensitivity Detection Reagent (Part B) is provided in the CBA Mouse Enhanced Sensitivity Master Buffer Kit. The Enhanced Sensitivity Capture Bead and Detection Reagent (Part A) components of this flex set have been formulated to a 20× concentration to ensure product performance when multiplexed.
- Capable of measuring soluble analytes present in very low concentrations
- Must be used in conjunction with CBA Mouse Enhanced Sensitivity Master Buffer Kit, flow cytometer, and the FCAP Array Software
Specifications
Specifications
| Antigen | IL-12p70 |
| Cross Reactivity | Mouse |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Measures Mouse interleukin-12 p70, flow cytometry |
| Product Type | Mouse IL-12p70 enhanced sensitivity flex set (bead D7) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.