Learn More
Description
Includes
- Enhanced Sensitivity Capture Bead (1 vial)
- Detection Reagent (Part A) (1 vial)
- Standard (2 vials)
- Capable of measuring soluble analytes present in very low concentrations
- Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
- Top standard point for this assay is 200,000fg/mL when reconstituted according to instructions
Flow cytometry
Specifications
Specifications
| Antigen | IL-2 |
| Cross Reactivity | Mouse |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Liquid |
| For Use With (Application) | Measures Mouse interleukin-2, flow cytometry |
| Product Type | Mouse IL-2 enhanced sensitivity flex set (bead A5) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.