Learn More
BD Cytometric Bead Array Mouse IL-5 Enhanced Sensitivity Flex Set
Catalog No. BDB562234 Shop All BD Biosciences Products
Change view
Click to view available options
Mouse IL-5 Enhanced Sensitivity Flex Set; 100 tests
Catalog No.
BDB562234 Mouse IL-5 Enhanced Sensitivity Flex Set; 100 tests
1 options
Catalog No. BDB562234 Supplier BD Supplier No. 562234
Add to Cart
Add to Cart
Includes
Capture beads, detection reagent, standards.
For measuring mouse Interleukin-5 (IL-5) in serum and cell culture supernatant samples
  • Bead-based immunoassay
  • Bead A6
  • Also measures soluble analytes present in very low concentrations