Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array Mouse IL-5 Flex Set Standard

Catalog No. BDB558478 Shop All BD Biosciences Products
Change view
Click to view available options
Kit Size:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Kit Size
BDB558478 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558478 Supplier BD Supplier No. 558478

missing translation for 'validMainframeMsg'

Lyophilized Mouse IL-5

  • Lyophilized in an aqueous buffered solution containing BSA and ProClin™ 150
  • Protein concentration of 25,000pg/ml (when reconstituted in 0.4ml Assay Diluent)
  • Regulatory Status: RUO
  • Recombinant

Flow cytometry, controls

Order Info

100 tests.

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffered Solution containing BSA and ProClin™ 150
Includes Technical data sheet
Kit Size 100 Tests
Product Type Cytometric Ab Control Standards
For Use With (Application) Flow cytometry
Regulatory Status RUO
Protein Concentration 25000pg/mL
Storage Requirements 4°C
Volume (Metric) 0.2mL
Species Murine
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.