Learn More
Description
Includes
Capture beads, detection reagent, standards.
- Bead-based immunoassays
- Bead B4
- Also measures soluble analytes present in very low concentrations
Specifications
Specifications
| Antigen | IL-6 |
| Cross Reactivity | Mouse |
| Includes | 1 Vial capture bead, 1 vial detection reagent, 2 vials standard (lyophilized) |
| Quantity | 100 Tests |
| Sample Type | Serum, Cell Culture Supernatant |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Measures Mouse interleukin-6, flow cytometry |
| Product Type | Mouse IL-6 enhanced sensitivity flex set (bead B4) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.