Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array Mouse Th1/Th2 Cytokine Standard

Catalog No. BDB552967 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552967 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552967 Supplier BD Supplier No. 552967
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed for use as a standard control with the Cytometric Bead Array (CBA) Mouse Th1/Th2 Cytokine Kit

  • Contains a mixture of individually lyophilized recombinant proteins made up of recombinant mouse IL-2, IL-4, IL-5, IFN-γ, and TNF
  • Lyophilized and should be reconstituted and serially diluted before mixing with CBA capture beads and PE detection reagents

TRUSTED_SUSTAINABILITY

Specifications

Buffer Aqueous Buffered Solution containing BSA and ProClin™ 150
Includes Manual
Product Type Cytometric Ab Control Standards
Formulation Mixture of individually lyophilized recombinant proteins made up of recombinant mouse interleukin (IL)-2, IL-4, IL-5, Interferon-γ (IFN-γ) and tumor necrosis factor (TNF)
For Use With (Application) Flow cytometry
pH 7.2
Regulatory Status RUO
Storage Requirements 4°C
Form Lyophilized
Quantity 1 Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.