Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array Th1/Th2 Cytokine Lyophilized Standard

Catalog No. BDB551810 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551810 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551810 Supplier BD Supplier No. 551810
Add to Cart
Add to Cart

Designed for use as a standard control with CBA Human Th1/Th2 Cytokine Kit and CBA Non-Human Primate Th1/Th2 Cytokine Kit

  • Contains a mixture of individually lyophilized recombinant proteins made up of human (IL)-2, IL-4, IL-5, IL-6, IL-10, Tumor Necrosis Factor (TNF), and IFN-γ
  • Lyophilized and should be reconstituted and serially diluted before mixing with CBA capture beads and PE detection reagents

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes Contains a mixture of individually lyophilized recombinant proteins made up of Human (IL)-2, IL-4, IL-5, IL-6, IL-10, Tumor Necrosis Factor (TNF) and IFN-g
Quantity 0.2 mL
Content And Storage 4°C
Form Lyophilized
For Use With (Application) Flow Cytometry
Product Type Human Th1/Th2 Cytokine lyophilized standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.