Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD D- Luciferin Sodium Salt

Catalog No. BDB556875 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556875 50 mg
BDB556876 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB556875 Supplier BD Supplier No. 556875
Add to Cart
Add to Cart

Includes

Sodium salt

Specific for firefly luciferase in the illustrated reactions

Purified by HPLC techniques to remove dehydroluciferin and L-Luciferin, both of which are competitive inhibitors of firefly luciferase. Form dehydroluciferin in presence of ultraviolet light and/or moisture

  • Synthetically made substrate
  • lyophilized, in monosodium salt form
  • Non-buffered solution
  • Reconstitute lyophilized D-Luciferin with deionized water

TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized
Kit Contents Sodium salt
Type D-Luciferin
Quantity 50 mg
Content And Storage -20°C
For Use With (Application) Luciferase assay

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.