Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD DAB Substrate Kit

Catalog No. BDB550880 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550880 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550880 Supplier BD Supplier No. 550880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

DAB buffer (50mL) is a solution of citric acid and H2O2. DAB chromagen (2.5mL) is a solution of 3-3 Diaminobenzidine(1.74% w/v) and HCl.

For immunoassay protocols

  • Substrate for enzyme peroxidase
  • Chromogen
  • Produces brown precipitate insoluble in alcohol

Immunohistochemistry-frozen/paraffin

TRUSTED_SUSTAINABILITY

Specifications

Chromagen Quantity 2.5mL
Chromagen Type DAB
For Use With (Application) Immunohistochemistry frozen or paraffin
Quantity 1 kit
Substrate Type DAB Substrate
Includes 50mL DAB buffer is a solution of citric acid and H2O2, 2.5mL DAB chromagen, is a solution of 1.74% w/v 3-3 Diaminobenzidine and HCl
pH 7.2
Storage Requirements 4°C

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.