Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difc™o Dehydrated Culture Media Additive: Supplement VX w/Reconstituting Fluid

Catalog No. DF3354603 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF3354-60-3 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF3354-60-3 Supplier BD Supplier No. 233541
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sterile concentrate of essential growth factors, V and X

TRUSTED_SUSTAINABILITY

Specifications

Additive Supplement VX with Reconstituting Fluid
Form Liquid
For Use With (Application) Supplementing media to culture fastidious microorganisms
Packaging Type Vial
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.