Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Antimicrobic Vial A Chlortetracycline

Catalog No. DF3333609 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF3333-60-9 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
ö
Catalog No. DF3333-60-9 Supplier BD Supplier No. 233331
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
ö

Specifications

Certifications/Compliance SMWW
Content And Storage 2°C to 8°C
Additive Antimicrobic vial A Chlortetracycline
Form Powder
For Use With (Application) Preparation of Bacto Cooke Rose Bengal Agar
Packaging Type Vial
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.