Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Antimicrobic Vial K Kanamycin

Catalog No. DF3339603 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Additive:
Antimicrobic vial K Kanamycin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Additive
DF3339-60-3 Antimicrobic vial K Kanamycin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF3339-60-3 Supplier BD Supplier No. 233391
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Certifications/Compliance AOAC, BAM, COMPF, ISO
Content And Storage 2°C to 8°C
Additive Antimicrobic vial K Kanamycin
Form Powder
For Use With (Application) SFP Agar base
Packaging Type Vial
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.