Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Bordetella spp. Antigen and Antisera

Catalog No. DF2585562 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF2585-56-2 5 mL
DF2310-50-0 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2585-56-2 Supplier BD Supplier No. 225851
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Quick reactive Gold standard antisera-typing panels for Bordetella Pertusis antigen and Bordetella Parapertusis antisera

  • Product type:     Antigens and Antisera
  • Groupings: A through B products
Bordetella Pertussis Antigen
  • Sub type: Quality control antigens
  • Type: Antigens
  • Quantity: 5mL
  • Shelf Life: 730 days
  • Positive Control for Bordetella pertussis antiserum
Bordetella Parapertussis Antiserum
  • Subtype: Bordetella Antisera
  • Type: Antisera
  • Lyophilized, rehydrate to 1mL
  • Shelf Life: 965 days
  • Rabbit serum for slide agglutination

Certifications

CE marked

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Type Antisera
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.