Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Culture Media Additives: Egg Yolk Enrichment 50%

Catalog No. B215329 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Egg Yolk Enrichment 50%; 10 x 10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B215329 Egg Yolk Enrichment 50%; 10 x 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B215329 Supplier BD Supplier No. 215329
Add to Cart
Add to Cart

To enrich media for cultivating and isolating large variety of microorganisms and tissue cells

Egg yolk enrichment 50%

  • For use in SFP Agar Base and McClung Toabe Agar Base for isolation of Clostridium species based on their lecithinase activity

TRUSTED_SUSTAINABILITY

Specifications

Concentration 50%
Content And Storage 2°C to 8°C
Additive Egg Yolk Enrichment
For Use With (Application) For use in Difco™ SFP agar base, Difco™ McClung Toabe Agar base for isolation and identification of clostridium species and for enrichment of media
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.