Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Francisella tularensis Antigen and Antisera

Catalog No. 14911128 Shop All Waters Corporation Products
Change view
Click to view available options
Type:
Francisella tularensis Antigen
Francisella tularensis Antisera
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-911-128 Francisella tularensis Antisera
14-911-127 Francisella tularensis Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-911-128 Supplier BD Supplier No. 241116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Febrile Control Antisera are used in agglutination tests in conjunction with Febrile Antigens as positive controls

  • Aids in diagnosis of certain febrile diseases such as salmonellosis, brucellosis and rickettsial diseases
  • Francisella Tularensis Antigen contains 0.5% formaldehyde in place of the phenol
  • Febrile Control Antisera are preparations of rabbit antiserum with 30% to 50% glycerin as preservative
  • Available in vials of 5mL

For in vitro Diagnostic Use.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE Marked
Content And Storage 2°C to 8°C
Type Francisella tularensis Antisera
Quantity 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.