Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ HYcheck™ Contact Slides: D/E Neutralizing Agar

Catalog No. DF0886082 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF-0886-08-2 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF-0886-08-2 Supplier BD Supplier No. 290001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to assess the microbiological contamination of surfaces or fluids.

  • Designed for environmental testing
  • Double sided
  • Hinged paddle bends for easy sampling

Order Info

Ten units per pack.

TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Content And Storage 2°C to 8°C
Detects Microbiological Contamination
Form Solid
Manufacturing Origin United States
Media Type D/E Neutralizing Agar
Product Type Contact Slide
Formulation Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 10g/L, Sodium Thioglycollate 1g/L, Sodium Thiosulfate 6g/L, Sodium Bisulfite 2.5g/L, Polysorbate 80 5g/L, Lecithin 7g/L, Bromcresol Purple 0.02g/L, Agar 15g/L
For Use With (Application) Used for assess the microbiological contamination of surfaces or fluids
Material Agar
No. per Pack 10/Pk.
Packaging Type Slide
Quantity 10 U
Specimen Type Surfaces, Fluids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.