Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ HYcheck™ Contact Slides: Yeasts and Molds with TTC

Catalog No. DF0886088 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF-0886-08-8 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF-0886-08-8 Supplier BD Supplier No. 290007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

To assess microbiological contamination of surfaces or fluids. BD Difco™ HYcheck™ Contact Slides: Yeasts and Molds with TTC are designed for environmental testing.

  • Designed for environmental testing
  • Double sided with Tryptic Soy Agar with 0.01% TTC and Rose Bengal Chloramphenicol Agar
  • Hinged paddle bends for easy sampling

Order Info

Ten units per pack.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Additive TTC
Detects Microbiological Contamination
Form Solid
Manufacturing Origin United States
Media Type Tryptic Soy Agar, Rose Bengal Chloramphenicol Agar
Product Type Contact Slide
Sterility Sterile
Target Organism Class Fungi
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L, 0.01% TTC, Papaic digest of Soybean Meal 5g/L, Dextrose 10g/L, Monopotassium Phosphate 1g/L, Magnesium Sulfate 0.5g/L, Rose Bengal 0.05g/L, Agar 15g/L, Chloramphenicol 0.05g/L
For Use With (Application) To assess microbiological contamination of surfaces or fluids
No. per Pack 10/Pk.
Packaging Type Slide
Quantity 10 U
Specimen Type Surfaces, Fluids
Protein Source Soy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.