Learn More
Description
Applications:
Used with Legionella Agar base for isolation of Legionella
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Additive | Legionella Agar Enrichment |
| Form | Liquid |
| For Use With (Application) | Legionella Agar base for the isolation of Legionella |
| Packaging Type | Bottle |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.