Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Modified TSB Prepared Media with Casamino Acids

Catalog No. 22350790 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Pail; Quantity: 10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-350-790 Pail; Quantity: 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-350-790 Supplier BD Supplier No. 215235
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used in qualitative procedures for the sterility test and for the enrichment and cultivation of aerobic microorganisms

  • Used in qualitative procedures for the sterility test.
  • Used for the enrichment and cultivation of aerobic microorganisms.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014, BAM, CCAM, CMPH2, COMPF, EP, JP, MCM9, USP
Detects E. coli , Aerobic Microorganisms
Form Solid
Format Powder, Pail
Manufacturing Origin United States
Media Type Modified Trypticase Soy Broth with Casamino Acids
For Use With (Application) Sterility testing and enrichment and cultivation of aerobic microorganisms
No. per Pack 1
Packaging Type Tube
Quantity 10 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.