Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Novobiocin Antimicrobic Supplement

Catalog No. DF319760 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF3197-60 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF3197-60 Supplier BD Supplier No. 231971
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applications:

Used with Brilliant green Agar, modified EC medium and Rappaport Vassiliadis (MSRV) semisolid modification

Adjusted and/or supplemented as required to meet performance criteria.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Novobiocin antimicrobic Supplement
Form Liquid
For Use With (Application) Brilliant green Agar, modified EC medium and Rappaport Vassiliadis (MSRV) semisolid modification
Packaging Type Bottle
Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.