Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ XLT4 Agar Supplement

Catalog No. DF0353726 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
XLT4 Agar Supplement; BD Difco; 100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
DF0353-72-6 XLT4 Agar Supplement; BD Difco; 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DF0353-72-6 Supplier BD Supplier No. 235310

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For isolation of non-typhi Salmonella

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 15°C to 30°C
Additive XLT4 Agar Supplement
Form Liquid
For Use With (Application) Isolating non-typhi Salmonella
Packaging Type Bottle
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.