Learn More
BD Directigen™ Group B Strep Test Kit
Catalog No. L855460 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
90 Tests
Catalog No. Quantity
L855460 90 Tests
1 options
This item is not returnable. View return policy
Catalog No. L855460 Supplier BD Supplier No. 255460
This item is not returnable. View return policy
Direct qualitative detection of antigens to Group B Streptococcus in cerebrospinal fluid (CSF) and serum. Provides confirmation and serogrouping capabilities from suspected colonies of Group B Streptococcus.
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity High, Moderate
Sample Type Blood Culture, CSF, Serum, Urine
Type Group B Streptococcus Test Kit
Includes Bottle group B Strep Antibody 1mL, R1, one bottle positive control 1.5mL, one bottle negative control 1.5mL, one bottle control Latex reagent A stirrers 0.5mL, Plastic
Quantity 90 Tests
Test Time 15 to 20 min.