Learn More
BD Directigen™ RSV Test Kit
Catalog No. L853040 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
40 Tests
Catalog No. Quantity
L853040 40 Tests
1 options
Catalog No. L853040 Supplier BD Supplier No. 253040
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Includes
Complete kit includes workstation, procedural chart, reagents, BD ColorPAC™ devices, tubes, tips, and positive and negative controls.
Ideal as a STAT test to provide rapid and relevant information
  • CLIA Complexity: Moderate
  • Ready-to-use in vitro enzyme immunoassay (EIA) membrane test
  • For the rapid qualitative detection of respiratory syncytial virus (RSV) antigen from nasopharyngeal specimens
  • Results in less than 10 minutes
  • Reactivity determined by a color readout
  • Storing kit at room temperature allows for a true STAT result
TRUSTED_SUSTAINABILITY
Specifications
Quantity 40 Tests