BD Disposable Syringes with Luer-Lok™ Tips

Unique integrated BD Luer-Lok™ tip syringe with clear barrel.   BD Medical™ Disposable Syringes with Luer-Lok™ Tips are sterile and disposable.

Manufacturer: BD 309646

 View more versions of this product

Catalog No. 14-829-45

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Disposable Yes
    Volume (Metric) 5mL
    Connection Type Luer Tip
    Latex Free Yes
    Termination Luer-lock
    Capacity (Metric) 5.0mL
    Sterile Yes
    Type Syringe

    • Sterile syringe for single use
    • Without needle
    • Clear barrel (polycarbonate or polypropylene) with bold scale markings
    • Tapered plunger rod for ease of aspiration
    • Positive plunger rod stop
    • Added BD Luer-Lok™ thread for increased secure connection
    • DNR (Latex) free
    • Shelf life: 1825 days

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit