Learn More
Description
Includes
Capture Antibody: Anti-mouse TNF-α, Detection Antibody: Biotinylated anti-mouse TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP) (250X), Standard: Recombinant mouse TNF-α (lyophilized)
- Flexible assay
- Calibrated against purified Baculovirus-expressed recombinant mouse TNF
ELISA
Specifications
Specifications
| Concentration | 15.6 to 1000pg/mL |
| Product Type | Mouse TNF ELISA set II |
| Sufficient For | 20 96-well Plates |
| For Use With (Application) | To develop ELISA for natural or recombinant Mouse TNF in Serum, Plasma and cell culture supernatants |
| Kit Contents | Biotin anti-Mouse TNF (250X) (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse TNF (standard), Purified anti-Mouse TNF (250X) (capture) antibody, Streptavidin-HRP Conjugate (250X) |
| Quantity | 20 Plates |
| Storage Requirements | 2°C to 8°C |
Research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.