Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD ELISA Set

Catalog No. BDB558315 Shop All BD Biosciences Products
Change view
Click to view available options
Sufficient For:
5 Plates of 96-well
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
BDB558315 5 Plates of 96-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558315 Supplier BD Supplier No. 558315
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Capture Antibody: Purified anti-human C5b-9 (250X)
  • Detection Antibody: Biotin anti-human C5b-9 (250X)
  • Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP) (250X)
  • Standards: Human C5b-9 Standard (5 vials)

To develop enzyme-linked immunosorbent assays for natural C5b-9 in serum, plasma, and other biological samples

  • Allows flexible assay design to fit individual laboratory needs
  • Immunoassay is calibrated against purified Cobra Venom-Factor activated human serum
  • Immunoassay with different sensitivity and dynamic range

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Product Type Human C5b-9 ELISA set
Sufficient For 5 Plates of 96-well
For Use With (Application) To develop ELISA for natural C5b-9 in Serum, Plasma and other biological samples
Kit Contents Biotin anti-Human C5b-9 (250X) (detection) Antibody, Human C5b-9 Standard (5 vials) (standard), Instruction/analysis certificate, Purified anti-Human C5b-9 (250X) (capture) Antibody, Streptavidin-HRP Conjugate (250X)
Quantity 5 Plates
Storage Requirements 4°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.