Learn More
BD Cell Analysis 3P Lysing Solution 10X Concentrate 100ml
Catalog No. 50620715
Encompass
BD FACS Lysing Solution, 100mL. IVD/10x-500 tests. pH value ranges from 7.2-7.6. Boiling point: -21C. Flash point: 146C. Ignition temperature: 225C. Product is not self igniting.
Catalog No. 50-620-715 Supplier BD Cell Analysis 3P Supplier No. 349202
BD FACS Lysing Solution, 100mL. IVD/10x-500 tests. pH value ranges from 7.2-7.6. Boiling point: -21C. Flash point: 146C. Ignition temperature: 225C. Product is not self igniting.