Learn More
BD Cell Analysis 3P Permeabilizing Solution 2   (RUO (GMP))
Catalog No. 50620449
Encompass
BD FACS™ Permeabilizing Solution 2 (10X) is a premixed concentrate formulated specifically for use in flow cytometry. It is intended for the permeabilization of lymphocyte membranes prior to intracellular immunofluorescence staining with monoclonal antibodies. This formulation is designed for antigen-specific activation applications.
Catalog No. 50-620-449 Supplier BD Cell Analysis 3P Supplier No. 340973
BD FACS™ Permeabilizing Solution 2 (10X) is a premixed concentrate formulated specifically for use in flow cytometry. It is intended for the permeabilization of lymphocyte membranes prior to intracellular immunofluorescence staining with monoclonal antibodies. This formulation is designed for antigen-specific activation applications.